Sodium dihydrogen phosphate dihydrate ≥98.0% USP

Supplier: Spectrum Chemical Mfg. Corp.

S1930-12KGBL S1930-1KG S1930-500GM S1930-50KGBL
75812-398EA 1695.06 CAD
75812-398 75812-400 75812-402 75812-404
Sodium dihydrogen phosphate dihydrate ≥98.0% USP
Sodium dihydrogen phosphate dihydrate

Storage temperature - Keep tightly closed


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: NaH₂PO₄·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149209
CAS Number: 13472-35-0

Order Now

Specification Test Results

Assay (Anhydrous Basis) 98.0 - 103.0 %
pH 4.1 - 4.5
Water Determination 18.0 - 26.5 %
Insoluble Substances max. 0.2 %
Chloride max. 0.014 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR