Sodium dihydrogen phosphate, anhydrous ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic

SO187-12KG SO187-45KGBL SO187-25KGBL
CA700001-680EA 4851 CAD
CA700001-680 CA89079-020 75812-462
Sodium dihydrogen phosphate, anhydrous ≥98.0% USP
Sodium dihydrogen phosphate

Monobasic Sodium Phosphate, Anhydrous, USP is used as a buffering agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: NaH₂PO₄
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7

Order Now

Specification Test Results

Assay (NaH₂PO₄) 98.0 - 103.0 %
pH of a 5% Solution 4.1 - 4.5
Water max. 2.0 %
Al, Ca, & Related Elements To Pass Test
Arsenic (As) max. 8 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR