Potassium dihydrogen phosphate ≥98.0%, crystals NF

Supplier: Spectrum Chemical Mfg. Corp.

PO200-12KG PO200-2.5KG PO200-45KGBL PO200-500GM PO200-2.5KG-CS PO200-500GM-CS
CA700001-708EA 4559 CAD
CA700001-708 CA700001-710 CA700001-712 CA700001-714 75812-068 75812-070
Potassium dihydrogen phosphate ≥98.0%, crystals NF
Potassium dihydrogen phosphate

Monobasic Potassium Phosphate, Crystal, NF is used as a buffering agent. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: KH₂PO₄
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0

Order Now

Specification Test Results

Assay (KH₂PO₄) 98.0 - 100.5 %
Loss On Drying max. 1.0 %
Insoluble Substances max. 0.2 %
Fluoride max. 10 µg/g
Arsenic (As) max. 3 µg/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR