Mineral oil medium FCC, medium viscosity

Supplier: Spectrum Chemical Mfg. Corp.

M1194-20LT M1194-500MLGL
75811-608EA 1761.54 CAD
75811-608 75811-610
Mineral oil medium FCC, medium viscosity
Mineral oil medium

Mineral Oil, Medium Viscosity, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5

Order Now

Specification Test Results

Carbon Number at 5% Distillation Point 17 - 25
Specific Gravity 0.848 - 0.867
UV Absorbance (Polynuclear Hydrocarbons) To Pass Test
Viscosity @ 100°C 3 - 11 cSt
Lead (Pb) max. 1 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR