Methyl-4-hydroxybenzoate sodium salt ≥95.0% NF

Supplier: Spectrum Chemical Mfg. Corp.

M1277-125GM M1277-500GM
75811-638EA 416.63 CAD
75811-638 75811-640
Methyl-4-hydroxybenzoate sodium salt ≥95.0% NF
Methyl 4-hydroxybenzoate sodium salt

Methylparaben Sodium, NF is an excipient used as a preservative. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₈H₇NaO₃
Storage Temperature: Ambient
MDL Number: MFCD00016470
CAS Number: 5026-62-0

Order Now

Specification Test Results

Assay (Anhydrous Basis) 95.0 - 102.0 %
Completeness of Solution To Pass Test
pH of a 1 in 1000 Solution 9.5 - 10.5
Water max. 5.0 %
Chloride max. 0.035 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR