Methyl-4-hydroxybenzoate ≥98.0% NF
Supplier: SPECTRUM CHEMICAL MFG CORP CA
|
|
Synonyms:
Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
ME2163-2.5KG
ME163-100GM
ME163-50KGBL
CA11024-372EA
1529.99
CAD
CA11024-372
75811-724
75811-726
Methyl-4-hydroxybenzoate ≥98.0% NF
Methyl 4-hydroxybenzoate
Methylparaben, NF is an excipient used as a preservative. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
GHS hazard : Exclamation mark
Formula:
HOC₆H₄CO₂CH₃ MW: 152.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002352 CAS Number: 99-76-3 |
Specification Test Results
Assay (C₈H₈O₃) | 98.0 - 102.0 % |
Color of Solution | To Pass Test |
Acidity | To Pass Test |
Residue On Ignition | max. 0.1 % |
Related Substances | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...