Methyl-4-hydroxybenzoate ≥99.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

M1187-12KG M1187-50KGBL
75811-602EA 2476.18 CAD
75811-602 75811-604
Methyl-4-hydroxybenzoate ≥99.0% FCC
Methyl 4-hydroxybenzoate

Methylparaben, FCC is used as a food preservative and has the E number E218. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: HOC₆H₄CO₂CH₃
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 100.5 %
Melting Range 125 - 128 °C
Acidity To Pass Test
Lead (Pb) max. 2 mg/kg
Loss On Drying max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR