Dichloromethane ≥99.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Methylene chloride

M1172-200LTBL M1172-20LT M1172-500MLGL
75811-594EA 6213.9 CAD
75811-594 75811-596 75811-598
Dichloromethane ≥99.0% FCC
Dichloromethane

Methylene Chloride, FCCis used to decaffeinate coffee and tea as well as to prepare extracts of hops and other flavorings. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark;Health hazard
UN1593

Formula: CH₂Cl₂
Density: 1.326
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
UN: 1593
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 %
Distilling Range 39.0 - 41.0 °C
Specific Gravity 1.318 -1.323
Acidity (As HCl) max. 10 mg/kg
Free Halogens To Pass Test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR