Methyl cellulose, 4,000 cP FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Cellulose methyl ether
M1164-100GM
M1164-12KG
M1164-2.5KG
M1164-45KGBL
M1164-500GM
75810-502EA
194.23
CAD
75810-502
75811-576
75811-578
75811-580
75811-582
Methyl cellulose, 4,000 cP FCC
Methyl cellulose
Methylcellulose, 4,000 cP, FCC is often used as a thickener and emulsifier.
All Spectrum Chemical FCC grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Storage Temperature:
Ambient |
MDL Number:
MFCD00081763 CAS Number: 9004-67-5 |
Specification Test Results
Assay (Methoxy Groups) | 27.5 - 31.5 % |
Viscosity of a 2G in 100 ML Soln | 3000 - 5600 cPs |
Lead (Pb) | max. 3 mg/kg |
Loss On Drying | max. 5.0 % |
Residue On Ignition | max. 1.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...