D-(-)-N-Methylglucamine ≥99.0% USP

Supplier: Spectrum Chemical Mfg. Corp.

M1122-100GM M1122-500GM M1122-2.5KG
95034-372EA 202.51 CAD
95034-372 95034-376 75811-562
D-(-)-N-Methylglucamine ≥99.0% USP
D(-)-N-Methylglucamine

Meglumine, USP is used as a functional excipient where it acts as a counterion to enhance API stability and solubility. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: H₃CNHCH₂(CH(OH))₄CH₂OH
Storage Temperature: Ambient
MDL Number: MFCD00004707
CAS Number: 6284-40-8

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 100.5 %
Completeness and Color of Solution To Pass Test
Melting Range 128 - 132 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR