D-(-)-Mannitol, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

M1128-12KG M1128-2.5KG M1128-500GM
75811-564EA 6709.55 CAD
75811-564 75811-566 75811-568
D-(-)-Mannitol, BiotechGrade
D(-)-Mannitol
Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Fine,White,Cryst
Assay (C₆H₁₄O₆; Dried Basis) 96.0 - 101.5 %
Appearance of Solution (2 G in 20 ML) Clear and Colorl
Melting Range 164 - 169°C
Specific Rotation, [A]D25 +137° to +145°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR