Manganese(II) sulfate monohydrate ≥98.0%, powder BP, FCC

Supplier: Spectrum Chemical Mfg. Corp.

MA164-12KG MA164-50KGBL
75811-708EA 2622.72 CAD
75811-708 75811-710
Manganese(II) sulfate monohydrate ≥98.0%, powder BP, FCC
Manganese (II) sulphate monohydrate

Manganese Sulfate, Monohydrate, Powder, FCC, BP - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Health hazard

Formula: MnSO₄·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149159
CAS Number: 10034-96-5
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay (MnSO₄[-H₂O) 98.0 - 102.0%
Assay (Ignited) 99.0 - 101.0%
Loss On Heating 10.0 - 12.0%
Appearance of Solution To Pass Test
Arsenic (As) max. 3 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR