DL-Malic acid ≥99.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: DL-Hydroxysuccinic acid, (±)-Malic acid, (±)-Hydroxy-1,4-butanedioic acid, (±)-Hydroxysuccinic acid, Hydroxybutanedioic acid, 2-Hydroxysuccinic acid

MA147-12KG MA147-45KGBL
75811-704EA 1691.07 CAD
75811-704 75811-706
DL-Malic acid ≥99.0% FCC
DL-Malic acid

Malic Acid, FCC is used as a source of extreme tartness. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: HOOCCH(OH)CH₂COOH
Storage Temperature: Ambient
MDL Number: MFCD00064212
CAS Number: 617-48-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₄H₆O₅) 99.0 - 100.5 %
Optical Rotation -0.10 to +0.10
Fumaric Acid max. 1.0 %
Lead (Pb) max. 2 mg/kg
Maleic Acid max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR