Maleic acid ≥99.0% NF

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

M1203-100GM M1203-12KG M1203-500GM M1203-50KGBL
75811-616EA 235.7 CAD
75811-616 75811-618 75811-622 75811-624
Maleic acid ≥99.0% NF
Maleic acid

Maleic Acid, NF is an acidifying agent excipient.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 


Caution: Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers. Store at room temperature.

Formula: C₄H₄O₄
MW: 116.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a1ea1f5bca5d5cf7fd40530f642f47db88908a&vid=1ac7f8cee2cd0433dbfb6e9271d41fe6ecc382c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.0 - 101.0 %
Color and Clarity of Solution To Pass Test
Fumaric Acid max. 1.5 %
Iron (Fe) max. 5 ppm
Heavy Metals max. 10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR