Magnesium sulfate heptahydrate 99,0 BP, Ph. Eur., USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: sulphuric acid magnesium salt heptahydrate, sulfuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

MA135-20KGBL MA135-45KGBL MA135-2.5KG MA135-500GM
CA700001-514EA 5067.99 CAD
CA700001-514 CA89078-868 95034-274 CA95034-276
Magnesium sulfate heptahydrate 99,0 BP, Ph. Eur., USP
Magnesium sulphate heptahydrate

MAGNESIUM SULFATE, HEPTAHYDRATE, USP, EP, BP is the commonly hydrated form of a highly hygroscopic material. Sterile solutions of magnesium sulfate heptahydrate, have been used as an electrolyte replenisher or anticonvulsant. Spectrum Chemical USP products, including all USP magnesium sulfate heptahydrate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical USP, EP, BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
Merck Index: 13,05715

Order Now

Specification Test Results

Assay (MgSO₄; Anhydrous Basis) 99.0 - 100.5 %
pH of a 5% Solution @ 25°C 5.0 - 9.2
Loss On Ignition 48.0 - 52.0 %
Appearance of Solution To Pass Test
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR