Magnesium oxide heavy ≥96.0%, powder USP

Supplier: Spectrum Chemical Mfg. Corp.

MA125-13 MA125-25KGBL
CA11024-366EA 1694 CAD
CA11024-366 75811-698
Magnesium oxide heavy ≥96.0%, powder USP
Magnesium oxide

Magnesium Oxide, Heavy Powder, USP has many uses, but from a pharmaceutical perspective it is used primarily as a mineral supplement and a tablet and capsule diluent. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: MgO
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4

Order Now

Specification Test Results

Assay 96.0 - 100.5%
Bulk Density 0.25 - 0.5 G/CC
Loss On Ignition max. 10.0%
Free Alkali & Soluble Salts To Pass Test
Acid-Insoluble Substances max. 0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR