Magnesium chloride hexahydrate 99,0, flake FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Magnesium dichloride

MA111-12KG MA111-50KGBL
75811-694EA 3129.82 CAD
75811-694 75811-696
Magnesium chloride hexahydrate 99,0, flake FCC
Magnesium chloride hexahydrate

Magnesium Chloride, Hexahydrate, Flake, FCC is used as a nutritional supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Boiling Pt: 1412
Melting Pt: 117
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
Merck Index: 13,05684

Order Now

Specification Test Results

Assay (MgCl₂[-6H₂O) 99.0 - 105.0 %
Ammonium (NH₄) max. 0.005 %
Lead (Pb) max. 4 mg/kg
Sulfate max. 0.03 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR