Magnesium chloride hexahydrate ≥98.0%, crystals USP, BP, Ph. Eur.

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Magnesium dichloride

MA110-12KG MA110-50KGBL MA110-500GMPL
CA700001-064EA 6162 CAD
CA700001-064 CA89078-852 CA10018-686
Magnesium chloride hexahydrate ≥98.0%, crystals USP, BP, Ph. Eur.
Magnesium chloride hexahydrate

Magnesium Chloride, Hexahydrate, Crystal, USP, EP, BP is used as a nutritional supplement. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: MgCl₂·6H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6

Order Now

Specification Test Results

Assay 98.0 - 101.0%
pH of a 5% Solution @ 25°C 4.5 - 7.0
Water 51.0 - 55.0%
Insoluble Matter max. 0.005%
Sulfate max. 0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR