Magnesium carbonate heavy FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Magnesium carbonate hydroxide, Magnesium carbonate basic, heavy
Magnesium Carbonate, Heavy, FCC is used as a nutritional supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
CH₂Mg₂O₅ MW: 142.63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011107 CAS Number: 39409-82-0 |
Specification Test Results
Assay (MgO) | 40.0 - 43.5 % |
Acid-Insoluble Substances | max. 0.05 % |
Calcium Oxide (CaO) | max. 0.6 % |
Lead (Pb) | max. 2 mg/kg |
Soluble Salts | max. 1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...