L(-)-Tyrosine ≥98.5% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH

T1150-25GM T1150-5KG
75812-542EA 145.76 CAD
75812-542 75812-544
L(-)-Tyrosine ≥98.5% FCC
L(-)-Tyrosine

L-Tyrosine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351
Melting Pt: 285
Storage Temperature: Ambient
MDL Number: MFCD00002606
CAS Number: 60-18-4
Merck Index: 13,09907

Order Now

Specification Test Results

Assay (Dried Basis) 98.5 - 101.5 %
Optical Rotation -10.0 To-11.0
Lead (Pb) max. 5 mg/kg
Loss On Drying max. 0.3 %
Residue On Ignition max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR