L(+)-Lysine monohydrochloride ≥98.5% FCC

Supplier: Spectrum Chemical Mfg. Corp.

L1143-1KG L1143-50KGBL
75810-496EA 615.96 CAD
75810-496 75811-492
L(+)-Lysine monohydrochloride ≥98.5% FCC
L(+)-Lysine monohydrochloride

L-Lysine Monohydrochloride, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Storage Temperature: Ambient
MDL Number: MFCD00064564
CAS Number: 657-27-2

Order Now

Specification Test Results

Assay (C₆H₁₄N₂O₂.HCl; Dried Basis) 98.5 - 101.5 %
Optical Rotation, @ 20D +20.3 to +21.5
Lead max. 5 mg/kg
Loss On Drying max. 1.0 %
Residue On Ignition max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR