Lime oil FCC, distilled

Supplier: SPECTRUM CHEMICAL MFG CORP

L1086-15KGBL L1086-500MLGL
75811-484EA 3354.89 CAD
75811-484 75811-486
Lime oil FCC, distilled
Lime oil

Lime Oil, Distilled, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Flame
UN1197

Density: 0.861
Storage Temperature: Ambient
MDL Number: MFCD00147443
CAS Number: 8008-26-2
UN: 1197
ADR: 3,III

Order Now

Specification Test Results

Assay (Aldehydes As Citral) 0.5-2.5 %
Angular Rotation +34 to +47
Refractive Index @ 20°C 1.474-1.477
Solubility in Alcohol To Pass Test
Specific Gravity @ 25°C 0.855-0.863


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR