L(+)-Histidine ≥98.5% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: L-3-Imidazol-4-ylalanine(S)-(+)-Histidine(S)-2-Amino-3-(4-imidazolyl)propionic acidNSC 137773L-HistidineH-His-OH

H1015-5KG H1015-1KG H1015-25GM H1015-100GM
75811-302EA 10381.33 CAD
75811-302 95033-960 95033-962 95033-964
L(+)-Histidine ≥98.5% FCC
L(+)-Histidine

L-Histidine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064315
CAS Number: 71-00-1
https://ca.vwr.com/stibo/web/std.lang.all/98/29/2299829.jpg

Order Now

Specification Test Results

Assay (Dried Basis) 98.5 - 101.5 %
Optical Rotation +11.5 to +13.5
Lead (Pb) max. 5 mg/kg
Loss On Drying max. 0.2 %
Residue On Ignition max. 0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR