L(+)-Glutamic acid ≥98.5% FCC

Supplier: Spectrum Chemical Mfg. Corp.

G1036-100GM G1036-12KG
95033-944EA 219.18 CAD
95033-944 75811-222
L(+)-Glutamic acid ≥98.5% FCC
L(+)-Glutamic acid

L-Glutamic Acid, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₅H₉No₄; Dried Basis) 98.5 - 101.5 %
Optical Rotation, @ 20D +31.5 to +32.5
Lead max. 5 mg/kg
Loss On Drying max. 0.1 %
Residue On Ignition max. 0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR