L(+)-Cysteine hydrochloride monohydrate ≥98.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate(R)-(+)-Cysteine hydrochloride monohydrateL-Cysteine HCI MonohydrateCysteine Hydrochloride monohydrateH-Cys-OH.HCl.H2OCysteine Hydrochloride, Monohydrate

C1472-1KG C1472-12KGBL C1472-25GM C1472-500GM C1472-100GM
75810-820EA 6722.79 CAD
75810-820 95033-870 95033-872 95033-874 95033-876
L(+)-Cysteine hydrochloride monohydrate ≥98.0% FCC
L(+)-Cysteine hydrochloride monohydrate

L-Cysteine Monohydrochloride, Monohydrate, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
MW: 175.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065606
CAS Number: 7048-04-6
https://ca.vwr.com/stibo/web/std.lang.all/58/37/2345837.jpg

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 101.5 %
Optical Rotation +5.0 to +8.0
Loss On Drying 8.0 - 12.0 %
Residue On Ignition max. 0.1 %
Lead (Pb) max. 5 mg/kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR