D-Lactose monohydrate, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: D-(+)-Lactose monohydrate

L1037-12KG L1037-2.5KG L1037-45KGBL L1037-500GM
75811-466EA 2508.01 CAD
75811-466 75811-468 75811-470 75811-472
D-Lactose monohydrate, BiotechGrade
D-Lactose monohydrate

Lactose, Monohydrate, BiotechGrade, also known as milk sugar, is a disaccharide sugar and is added to pills as a filler because of its physical properties. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Formula: C₁₂H₂₂O₁₁·1H₂O
MW: 360.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150747
CAS Number: 64044-51-5

Order Now

Specification Test Results

Appearance Fine,White Powde
Clarity and Color of Solution To Pass Test
Specific Rotation [A]D +54.4° to +55.9
Acidity / Alkalinity To Pass Test
Water 4.5 - 5.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR