Lactobionic acid ≥98.0% Ph. Eur.

Supplier: Spectrum Chemical Mfg. Corp.

L1604-500GM L1604-5KGBL
75811-512EA 1611.81 CAD
75811-512 75811-514
Lactobionic acid ≥98.0% Ph. Eur.
Lactobionic acid

Lactobionic Acid, NF, EP is also known as Galactosylgluconic acid. It is a food additive used as a stabilizer and in the pharmaceutical industry as an excipient for formulation. The EP grade indicates it is grade suitable for personal care, cosmetic and pharmaceutical applications.


All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₂H₂₂O₁₂
Storage Temperature: Ambient
MDL Number: MFCD00078147
CAS Number: 96-82-2

Order Now

Specification Test Results

Assay (Anhydrous Basis) 98.0 - 102.0 %
Appearance of Solution To Pass Test
Specific Optical Rotation +23.0 to +29.0
Reducing Sugars (As Glucose) max. 0.2 %
Heavy Metals max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR