Lactic acid racemate USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

L1010-20LTPL L1010-4LTPL L1010-4LTPL-CS
CA700006-076EA 7048 CAD
CA700006-076 75811-462 75811-464
Lactic acid racemate USP
Lactic acid

LACTIC ACID, RACEMIC, USP, also known as milk acid and is used as a sanitizer in pharmaceutical production areas. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. Storage temperature - HYGROSCOPIC: Keep tightly closed.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Density: 1.209
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Assay (C₃H₆O₃) 88.0 - 92.0%
Specific Rotation -0.05° to +0.05
Residue On Ignition max. 0.05%
Sugars To Pass Test
Chloride To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR