2-Propanol 70% ≥70% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

IS120-1LTGL-CS IS120-4LTGL-CS IS120-4LTPL-CS IS120-500ML-CS
75811-438CS 941.19 CAD
75811-438 75811-440 75811-442 75811-444
2-Propanol 70% ≥70% USP
2-Propanol

Isopropyl Rubbing Alcohol, 70 Percent, USP is a colorless and flammable chemical compound also known as Rubbing Alcohol. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: (CH₃)₂CHOH
Density: 0.785
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₃H₈O) 68.0 - 72.0 %
Acidity To Pass Test
Specific Gravity @ 20°C 0.872 - 0.883
Nonvolatile Residue max. 0.01 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR