2-Propanol 99 JP, USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

IS115-1LTGL-CS IS115-4LTGL-CS IS115-4LTPL-CS IS115-500ML-CS IS115-55GLBL
75811-428CS 1163.67 CAD
75811-428 75811-430 75811-432 75811-434 75811-436
2-Propanol 99 JP, USP
2-Propanol

Isopropyl Alcohol, 99 Percent, USP, JP has many uses, including as a pharmaceutical raw material for preparing USP Rubbing Alcohol for relief of muscle stiffness and an infection preventative for minor scrapes and burns.


GHS hazard : Flame;Exclamation mark;Health hazard UN1219

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.781
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3 II
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₃H₈O) 99.0% min.
Specific Gravity @ 25°C 0.783 - 0.787
Specific Gravity @ 20°C 0.785 - 0.788
Refractive Index @ 20°C 1.376 - 1.378
Acidity To Pass Test
Appearance Clear and colorless
Clarity of solution Solution is clear
Distilling range (Min. 94%) 81 - 83 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR