2-Propanol ≥99% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

I1056-1LT I1056-200LTBL I1056-20LT I1056-4LTGL I1056-4LTGL-CS I1056-4LTPL I1056-4LTPL-CS I1056-500ML-CS I1056-500MLGL
75811-402EA 263.94 CAD
75811-402 75811-404 75811-406 75811-408 75811-410 75811-412 75811-414 75811-416 75811-418 75811-420
2-Propanol ≥99% FCC
2-Propanol

Isopropyl Alcohol, 99 Percent, FCC is used to sanitize food processing equipment. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Flame;Exclamation mark;Health hazard
UN1219

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.781
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3 II
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.7 % min.
Distillation Range Within 1 °C
Acidity (As CH₃COOH) max. 10 mg/kg
Lead (Pb) max. 1 mg/kg
Nonvolatile Residue max. 10 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR