Iodine ≥99.8%, crystals USP, resublimed (DEA List I Chemical)

Supplier: SPECTRUM CHEMICAL MFG CORP

I1015-12KG I1015-500GM I1015-25GM I1015-500GM-CS I1015-2.5KG-CS I1015-125GM I1015-125GM-CS
75811-400EA 15146.91 CAD
75811-400 75812-748 75812-752 75812-754 75812-756 75812-758 75812-784 75812-786
Iodine ≥99.8%, crystals USP, resublimed (DEA List I Chemical)
Iodine

Iodine, Resublimed (DEA List I Chemical), Crystal, USP is a chemical element and it is often found as a contrast material in X-rays and water purification reagents.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: I₂
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3495
ADR: 86.1,III

Order Now

Specification Test Results

Assay (I₂) 99.8 - 100.5 %
Nonvolatile Residue max. 0.05 %
Chloride and Bromide (As Cl) max. 0.028 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR