Hydroxypropyl cellulose, 4,000 - 6,500 cP NF
Supplier: Spectrum Chemical Mfg. Corp.
HY121-2.5KG
HY121-125GM
HY121-12KG
HY121-500GM
95033-592EA
1751.51
CAD
95033-592
95033-590
700013-026
95033-594
Hydroxypropyl cellulose, 4,000 - 6,500 cP NF
Hydroxypropyl cellulose
Hydroxypropyl Cellulose, 4,000-6,500 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous formulations.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Storage Temperature:
Ambient |
MDL Number:
MFCD00132688 CAS Number: 9004-64-2 |
Specification Test Results
Assay (Hydroxypropoxy Groups) | 53.4 - 80.5 % |
Apparent Viscosity of a 2% Aqueous Soln. @25 C | As Reported |
pH of a 1% Solution | 5.0 - 8.0 |
Loss On Drying | max. 5.0% |
Silica or Other Anticaking Agents | max. 0.6% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...