Hydrogen peroxide solution 35% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP CA

HY115-4X4L HY115-4LTPL HY115-500MLPL HY115-200LTBL HY115-500ML-CS
CA10736-408EA 2538 CAD
CA10736-408 CA10746-648 CA10746-796 75811-396 75811-398
Hydrogen peroxide solution 35% FCC
Hydrogen peroxide

Hydrogen Peroxide, 35 Percent Solution, FCC is used as a disinfectant in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Flame over circle;Corrosion;Exclamation mark
UN2014

Formula: H₂O₂
MW: 34.01 g/mol
Density: 1.11
Storage Temperature: Refrigerator
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2014
ADR: 5.18,II

Order Now

Specification Test Results

Assay (H₂O₂) 35.0 % min.
Acidity (As H₂SO₄) max.0.03 %
Iron (Fe) max. 0.5 mg/kg
Lead (Pb) max. 4 mg/kg
Phosphate (Po₄) max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR