Hydrochloric acid 37% FCC

Supplier: Spectrum Chemical Mfg. Corp.

HY106-200LTBL HY106-20LT HY106-4LT HY106-500ML-CS HY106-500MLGL
75811-382EA 3440.61 CAD
75811-382 75811-384 75811-386 75811-388 75811-390 75811-392
Hydrochloric acid 37% FCC
Hydrochloric acid

Hydrochloric Acid, 37 Percent, FCC is used in food processing, or as a food additive to adjust the pH. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Corrosion;Exclamation mark
UN1789

Formula: HCl
Density: 1.200
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Assay 36.0 - 38.0 %
Color To Pass Test
Specific Gravity To Pass Test
Iron (Fe) max. 5 mg/kg
Lead (Pb) max. 1 mg/kg

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR