Gum arabic, Spray-Dried powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

GU113-12KG GU113-45KGBL
75811-292EA 3239.32 CAD
75811-292 75811-294
Gum arabic, Spray-Dried powder FCC
Gum arabic

Gum Arabic, Spray-Dried Powder, FCC is used in the food industry as an emulsifier, a confectionary coating and as a soluble dietary fiber. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
MDL Number: MFCD00081264
CAS Number: 9000-01-5

Order Now

Specification Test Results

Loss On Drying max. 15.0 %
Arsenic (As) max.3 mg/kg
Acid Insoluble Ash max. 0.5 %
Total Ash max. 4.0 %
Insoluble Matter max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR