Glycerine 99, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

G1433-20LTBL G1433-4LT G1433-500ML
75811-264EA 8992.65 CAD
75811-264 75811-266 75811-268
Glycerine 99, BiotechGrade
Glycerine

Glycerin, BiotechGrade, is a sugar alcohol (polyol) compound. An odorless, viscous and colorless liquid that has three hydroxyl groups that allow its solubility in water and give it it's hygroscopic nature. Used in the personal care and pharmaceutical industry for improving smoothness and lubrication it is also used as a solvent and preservative.

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.264
Flash Pt: 177
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Approx. 99 %
Appearance Clear Colorless
Ammonia Content (NH ) max. 0.05 %
Residue On Ignition max. 0.1 %
Chloride (Cl) max. 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR