Glycerine (natural) ≥99.0% USP, Ph. Eur., BP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

G1016-20LTPL G1016-1LT
CA700000-218EA 5842 CAD
CA700000-218 75811-218
Glycerine (natural) ≥99.0% USP, Ph. Eur., BP
Glycerine

Glycerin, Natural, USP, EP, BP is one of two USP grade Glycerins available from Spectrum.


All Spectrum Chemical USP, EP, BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Density: 1.264
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.0 - 101.0%
Specific Gravity @ 25°C 1.249 min.
Appearance of Solution To Pass Test
Refractive Index 1.470 - 1.475
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR