Ginger oil FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

GI115-100ML GI115-500ML
75811-282EA 675.21 CAD
75811-282 75811-284
Ginger oil FCC
Ginger oil

Ginger Oil, FCC is used in dietary supplements.


All Spectrum Chemical FCC dietary supplements are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
MDL Number: MFCD00147026
CAS Number: 8007-08-7

Order Now

Specification Test Results

Angular Rotation -28 To-47
Refractive Index @ 20°C 1.488-1.494
Specific Gravity @ 25°C 0.870-0.882
Saponification Value max. 20
Identification To Pass Test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR