Formic acid FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Aminic Acid, Methanoic acid

F1135-2.5LT-CS F1135-2.5LTGL F1135-500ML-CS
75811-136CS 2466.14 CAD
75811-136 75811-138 75811-140 75811-142 75811-144
Formic acid FCC
Formic acid

Formic Acid, FCC is the simplest carboxylic acid and one of its biggest uses is as a preservative and antibacterial agent in livestock feed. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Corrosion;Exclamation mark;Flame
UN1779

Formula: HCO₂H
Density: 1.220
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
UN: 1779
ADR: 8.3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (HCOOH) 85.0 % min.
Dilution Test To Pass Test
Acetic Acid (CH₃COOH) max. 0.4 %
Sulfate max. 0.004 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR