Formaldehyde 37% in aqueous solution USP
Supplier: Spectrum Chemical Mfg. Corp.
|
|
FO110-125ML
FO110-1LT
FO110-4LTPL
FO110-500MLGL
75811-210EA
122.85
CAD
75811-210
75811-212
75811-214
75811-216
Formaldehyde 37% in aqueous solution USP
Formaldehyde
Formaldehyde, 37 Percent, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Flame;Skull and crossbones;Corrosion;Exclamation mark;Health hazard
UN1198
Formula:
CH₂O Density: 1.08 Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 UN: 1198 ADR: 38,III |
Specification Test Results
Assay (Hcho) | 34.5% min. |
Acidity | To Pass Test |
Methanol | 9.0 - 15.0% |
Residual Solvents | To Pass Test |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...