Iron(II) sulfate heptahydrate ≥99.5%, granular USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ferrous sulphate heptahydrateIron vitriolFerrous sulfate heptahydrate

FE110-500GM FE110-50KGBL FE110-12KG FE110-2.5KG
75811-202EA 2391.98 CAD
75811-202 75811-204 95033-254 95033-256
Iron(II) sulfate heptahydrate ≥99.5%, granular USP
Iron (II) sulphate heptahydrate

Ferrous Sulfate, Heptahydrate, Granular, USP. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0

Order Now

Specification Test Results

Assay (As Heptahydrate) 99.5 - 104.5 %
Arsenic max.3 ppm
Lead max. 10 ppm
Mercury max. 3 Ug/G
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR