Iron(II) sulfate hydrate, dried powder FCC

Supplier: Spectrum Chemical Mfg. Corp.

F1063-12KG F1063-50KGBL
75811-132EA 2701.01 CAD
75811-132 75811-134
Iron(II) sulfate hydrate, dried powder FCC
Iron (II) sulphate hydrate

Ferrous Sulfate, Dried Powder, FCC is used as an iron fortification food supplement additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: FeSO₄·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149722
CAS Number: 13463-43-9

Order Now

Specification Test Results

Assay (FeSO₄) 86.0 - 89.0 %
Insoluble Substances max. 0.05 %
Lead (Pb) max. 2 mg/kg
Mercury max. 1 mg/kg
Identification To Pass Test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR