Erioglaucine disodium salt

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sodium 2-((4-(ethyl(3-sulfonatobenzyl)amino)phenyl)(4-(ethyl(3-sulfonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulfonate, FD and C Blue No. 1, Brilliant Blue FCF, Erioglaucin A, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)benzenesulphonate, Alphazurine FG, Erioglaucine sodium salt, CI 42090, Acid blue 9, Food Blue No.1

FD110-10KGBL FD110-25GM
75811-194EA 6351.52 CAD
75811-194 75811-196
Erioglaucine disodium salt
Erioglaucine disodium salt

FD and C Blue No. 1 | Non-graded FD and C Blue No. 1 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃₇H₃₄Na₂N₂O₉S₃
Storage Temperature: Ambient
MDL Number: MFCD00012141
CAS Number: 3844-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR