EDTA disodium salt dihydrate, BiotechGrade

Supplier: Spectrum Chemical Mfg. Corp.

E1058-12KGBL E1058-2.5KG E1058-2.5KG-CS E1058-45KGBL E1058-500GM E1058-500GM-CS
75811-038EA 4030.11 CAD
75811-038 75811-040 75811-042 75811-044 75811-046 75811-048
EDTA disodium salt dihydrate, BiotechGrade
EDTA disodium salt dihydrate

Edetate Disodium, Dihydrate, BiotechGrade is used in molecular biology applications to minimize metal ion contamination and prevent enzymatic activity. EDTA disodium dihydrate is routinely used in electrophoresis DNA and protein separation applications to chelate metal ions required for enzymatic activity that could potentially damage DNA and protein structure.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White,Crystallin
Odor Odorless
Assay (C₁₀H₁₄N₂Na₂O₈; Dried Basis) 99.0 - 101.0 %
pH of a 5% Solution @ 25°C 4.0 - 6.0
Appearance of a 5% Solution Clear & Colorles


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR