(R)-(+)-Limonene, for food industry

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (R)-(+)-Dipentene, (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexene, (R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexene, D(+)-Limonene, (R)-(+)-p-Mentha-1,8-diene, D-Limonene

L1599-500GM L1599-15KGBL L1599-2.5KG L1599-500GM
700002-382EA 273.92 CAD
700002-382 75811-508 75811-510 CA700002-382
(R)-(+)-Limonene, for food industry
(R)-(+)-Limonene

D-Limonene, Food Grade, also known as 4-Isopropenyl-1-methylcyclohexene, is used as a flavorant and as a fragrance in perfumery, aftershave lotions, bath products and other such products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available


GHS hazard : Flame;Health hazard;Exclamation mark
UN2052

Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 175.5
Melting Pt: -74
Density: 0.841
Flash Pt: 43
Storage Temperature: Ambient
MDL Number: MFCD00062991
CAS Number: 5989-27-5
UN: 2052
ADR: 3 III
Merck Index: 13,05515

Order Now

Specification Test Results

Appearance Transparent Liqu
Assay (GC) 95.0 - 97.5%
Specific Gravity @ 25°C 0.838 - 0.843
Refractive Index @ 20°C 1.471 - 1.474
Optical Rotation @ 25°C +96.0° to +104


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR