EDTA disodium salt dihydrate ≥99.0% FCC

Supplier: Spectrum Chemical Mfg. Corp.

E1001-12KG E1001-45KGBL
75811-010EA 3121.84 CAD
75811-010 75811-012
EDTA disodium salt dihydrate ≥99.0% FCC
EDTA disodium salt dihydrate

Disodium EDTA, Dihydrate, FCC is used as a preservative to inhibit discoloration. Spectrum Chemical manufactured FCC products, including all FCC Disodium products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₁₀H₁₄O₈N₂Na₂2H₂O) 99.0 - 101.0 %
pH of a 1 in 100 Solution 4.3 - 4.7
Calcium (Ca) To Pass Test
Lead (Pb) max. 10 mg/kg
Nitrilotriacetic Acid[N[(HOCOCH₂)₃N] To Pass Test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR