D-(+)-Glucose, anhydrous, granular, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

D1082-12KG D1082-2.5KG D1082-45KGBL D1082-500GM
75810-972EA 2254.82 CAD
75810-972 75810-974 75810-976 75810-978
D-(+)-Glucose, anhydrous, granular, BiotechGrade
D(+)-Glucose

Dextrose, Anhydrous, Granular, BiotechGrade is a simple monosaccharide sugar and is commonly referred to as glucose. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.


Spectrum offers highly pure reagents suitable for biochemical research and analysis.

Formula: C₆H₁₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White,Granular P
Odor Odorless
Appearance of a Solution (10 G in 15 ML) To Pass Test
Solubility (25 G in 50 ML) To Pass Test
Calcium (Ca) max. 200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR