Dehydrocholic acid ≥98.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

D1001-100GM D1001-1KGBL D1001-25GM D1001-500GM D1001-5KGBL
75810-956EA 1107.19 CAD
75810-956 75810-958 75810-960 75810-962 75810-964
Dehydrocholic acid ≥98.5% USP
Dehydrocholic acid

Dehydrocholic Acid, USP. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₂₄H₃₄O₅
Storage Temperature: Ambient
MDL Number: MFCD00066410
CAS Number: 81-23-2

Order Now

Specification Test Results

Assay (Dried Basis) 98.5 - 101.0 %
Melting Range (Max. 3 C Range) 231 - 242 °C
Specific Rotation [A]D +29.0 to +32.5
Microbial Limits To Pass Test
Loss On Drying max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR