Corn oil NF
Supplier: SPECTRUM CHEMICAL MFG CORP CA
CO136-4LTGL
CO136-25ML
CA700000-132EA
3075.49
CAD
CA700000-132
CA700000-134
75810-946
Corn oil NF
Corn oil
Corn Oil, NF its uses include as a solvent vehicle for topical preparations and as a glidant excipient for oral dosage tablets and capsules. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Storage Temperature:
Ambient |
MDL Number:
MFCD00130865 CAS Number: 8001-30-7 |
Specification Test Results
Fatty Acid Composition | To Pass Test |
Acid Value | max. 0.2 |
Peroxide Value | max. 10.0 |
Water Determination | max. 0.1 % |
Alkaline Impurities | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...